missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium sulfide nonahydrate, 98+%, ACS reagent, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC424420025

 View more versions of this product

Catalog No. AC424420025


 

Specifications Description & Specifications

Specifications

Sodium sulfide nonahydrate
sulfite and thiosulfate (as SO4): < 0.1%
Adhering Crystals Chunks
98+%
50ppm max.
240.18
01,1104; 11,492
Passes Test
Colorless or slightly yellow
1313-84-4
ACS Reagent Grade
≥98.0%
Na29H2O
Na2S·9H2O
MFCD00149184
15, 8810
Glass bottle
2.5kg

Chemical Identifiers

1313-84-4
Na29H2O
240.18
Sodium sulfide nonahydrate
ACS Reagent Grade
98+%
Na2S·9H2O
MFCD00149184
≥98.0%

Safety and Handling

SDS & Safety Documents

GHS P Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May be corrosive to metals.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Keep only in original container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

RTECSNumber : WE1925000