Learn More
Sodium sulfide nonahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
$46.21 - $697.26
Chemical Identifiers
CAS | 1313-84-4 |
---|---|
Molecular Formula | H18Na2O9S |
Molecular Weight (g/mol) | 240.17 |
MDL Number | MFCD00149184 |
InChI Key | ZGHLCBJZQLNUAZ-UHFFFAOYSA-N |
IUPAC Name | disodium nonahydrate sulfanediide |
SMILES | O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC424420250
|
Thermo Scientific Chemicals
424420250 |
25 g | Glass bottle |
Each for $46.21
|
|
||||
AC424425000
|
Thermo Scientific Chemicals
424425000 |
500 g | Glass Bottle |
Each for $211.12
|
|
||||
AC424420025
|
Thermo Scientific Chemicals
424420025 |
2.5 kg | Plastic bottle |
Each for $697.26
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-84-4 | |
240.17 | |
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] |
H18Na2O9S | |
MFCD00149184 | |
disodium nonahydrate sulfanediide |
Specifications
50.0°C | |
Colorless or Yellow | |
25 g | |
H18Na2O9S | |
MFCD00149184 | |
15, 8810 | |
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
disodium nonahydrate sulfanediide | |
240.18 | |
ACS Reagent | |
Sodium sulfide nonahydrate |
1313-84-4 | |
Adhering Crystals Chunks | |
98% min. (Iodimetry) | |
Na2S·9H2O | |
01,1104; 11,492 | |
Solubility in water: 180g/L (25°C). Other solubilities: slightly soluble in alcohol, insoluble in ether | |
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] | |
240.17 | |
≥98% | |
Glass bottle |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
Corrosiv
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
RTECSNumber : WE1925000
RUO – Research Use Only