Learn More
Sodium sulfide nonahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 424425000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium sulfide nonahydrate | |
1313-84-4 | |
Adhering Crystals Chunks | |
98+% | |
Na2S·9H2O | |
01,1104; 11,492 | |
Solubility in water: 180g/L (25°C). Other solubilities: slightly soluble in alcohol, insoluble in ether | |
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] | |
240.17 | |
≥98% | |
Glass Bottle |
50.0°C | |
Colorless or Yellow | |
500 g | |
H18Na2O9S | |
MFCD00149184 | |
15, 8810 | |
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
disodium nonahydrate sulfanediide | |
240.18 | |
ACS Reagent |
Chemical Identifiers
1313-84-4 | |
240.17 | |
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] |
H18Na2O9S | |
MFCD00149184 | |
disodium nonahydrate sulfanediide |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May be corrosive to metals.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Keep only in original container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RTECSNumber : WE1925000
RUO – Research Use Only