Learn More
Sodium sulfide hydrate, 60-64%, extra pure, flakes, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 133720025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium sulfide hydrate | |
27610-45-3 | |
2.5 kg | |
Na2S | |
MFCD00149183 | |
GRVFOGOEDUUMBP-UHFFFAOYSA-N | |
disodium;sulfanide;hydrate | |
45051681 | |
60 to 64% | |
Plastic bottle |
Yellow | |
Flakes and Chunks | |
60 to 64% (Na2S) (Iodimetry) | |
Na2S·xH2O | |
Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,insoluble in ether | |
[Na+].[Na+].[S--] | |
78.04 | |
78.04 | |
Extra Pure |
Chemical Identifiers
27610-45-3 | |
78.04 | |
GRVFOGOEDUUMBP-UHFFFAOYSA-N | |
[Na+].[Na+].[S--] |
Na2S | |
MFCD00149183 | |
45051681 |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
Corrosiv
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
RUO – Research Use Only