missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium sulfide, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 06512222

 View more versions of this product

Catalog No. AA6512222



Description

Description

In organic chemicals, dyes rayon, leather and paper, in hydrometallurgy of gold ore, sulfiding oxidized lead and copper ores, and as an analytical reagentSodium sulfide is used mainly in the pulp and paper industry in the Kraft process. It serves as an oxygen scavenger agent in water treatment. Other applications include ore flotation, oil recovery, making dyes and detergents. In the leather processing industry, it is utilized for liming operation as an unhairing agent. It is also used in the textile industry as a bleaching and desulfurizing agent and as a dechlorinating agent in chemical photography for toning black and white photographs as well as metals precipitant. It plays an important role as a sulfonation and sulfomethylation agent in the chemical industry and in the production of rubber chemicals, sulfur dyes and other chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium sulfide, anhydrous
1313-82-2
100 g
MFCD00003498
Soluble in water. Decomposes in acid. Slightly soluble in alcohol. Insoluble in etherSoluble in water. Slightly soluble in alcohol. Insoluble in ether.
[Na+].[Na+].[S--]
78.04
78.04
1.856 g/mL
950°C
Granules
Na2S
sodium sulfide, sodium sulfide na2s, disodium monosulfide, na2s, disodium sulphide, sodium sulfide van, hsdb 772, sodium sulfide, anhydrous dot, sodum sulfuret, na2-s
GRVFOGOEDUUMBP-UHFFFAOYSA-N
disodium;sulfanide
237873
Hygroscopic; Light sensitive

Chemical Identifiers

1313-82-2
78.04
GRVFOGOEDUUMBP-UHFFFAOYSA-N
237873
Na2S
MFCD00003498
sodium sulfide, sodium sulfide na2s, disodium monosulfide, na2s, disodium sulphide, sodium sulfide van, hsdb 772, sodium sulfide, anhydrous dot, sodum sulfuret, na2-s
[Na+].[Na+].[S--]
Safety and Handling

Safety and Handling

P234-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H301+H311-H314-H500

DOTInformation : Hazard Class: 4.2; Packaging Group: II

EINECSNumber : 215-211-5

RTECSNumber : WE1905000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only