missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium sulfide, anhydrous, Alfa Aesar™

Manufacturer:  Alfa Aesar 6512214

 View more versions of this product

Catalog No. AA6512214



Specifications

Specifications

Sodium sulfide, anhydrous
HNa2S+
sodium sulfide, sodium sulfide na2s, disodium monosulfide, na2s, disodium sulphide, sodium sulfide van, hsdb 772, sodium sulfide, anhydrous dot, sodum sulfuret, na2-s
[Na+].[Na+].[SH-]
79.048
78.04
1.856 g/mL
1313-82-2
MFCD00003498
VDQVEACBQKUUSU-UHFFFAOYSA-M
disodium;sulfanide
237873
Granular
25g

Chemical Identifiers

1313-82-2
79.048
VDQVEACBQKUUSU-UHFFFAOYSA-M
237873
[Na+].[Na+].[SH-]
HNa2S+
MFCD00003498
sodium sulfide, sodium sulfide na2s, disodium monosulfide, na2s, disodium sulphide, sodium sulfide van, hsdb 772, sodium sulfide, anhydrous dot, sodum sulfuret, na2-s
disodium;sulfanide
Safety and Handling

Safety and Handling

EINECSNumber : 215-211-5

SDS Documents