Please login to your online account to display your discounted pricing

Sodium Sulfide, 9-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$512.40 - $1,500.87

Chemical Identifiers

Chemical Name or Material Sodium Sulfide
CAS 1313-84-4
Name Note 9-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2S·9H2O
Formula Weight 240.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004243

 
avantor performance
391005
Poly Bottle 2.5kg Each for $1,500.87

02004244

 
avantor performance
391001
Wide Mouth Amber Glass Bottle 500g Each for $512.40
Description & Specifications

Specifications

Formula Weight 240.18
Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Chemical Name or Material Sodium Sulfide
Molecular Formula Na2S·9H2O
CAS 1313-84-4
Specific Gravity 1L = 1.43kg
Quantity 2.5kg
Name Note 9-Hydrate
Packaging Poly Bottle