Please login to your online account to display your discounted pricing

Sodium Sulfate USP, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sodium Sulfate
Name Note Anhydrous
CAS 7757-82-6
Purity Grade USP
Molecular Formula Na2SO4
Formula Weight 142.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK8028212

 
avantor performance
802806
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.04
Quantity 2.5kg
Purity Grade USP
Chemical Name or Material Sodium Sulfate
Name Note Anhydrous
Molecular Formula Na2SO4
Packaging Poly
CAS 7757-82-6