missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium sulfate, Puratronic™, 99.9955% (metals basis)

Manufacturer:  Alfa Aesar™ 1087422

 View more versions of this product

Catalog No. AA1087422



Specifications

Specifications

Sodium sulfate
(metals basis)
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
[O-]S(=O)(=O)[O-].[Na+].[Na+]
142.036
CHEBI:32149
Powder
Puratronic™
Odorless
Hygroscopic
100g
7757-82-6
Na2O4S
14,8680
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
142.04
99.9955%
2.680g/mL
1.484
884°C

Chemical Identifiers

7757-82-6
142.036
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
disodium;sulfate
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149
[O-]S(=O)(=O)[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

RTECSNumber : WE1650000

TSCA : TSCA

Documents