Please login to your online account to display your discounted pricing

Sodium Sulfate Decahydrate (Granular/USP), Fisher Chemical


Quantity 12kg
Packaging Poly Pail

Manufacturer: fisher scientific company  FLS41412

 View more versions of this product

Catalog No. S414-12

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 322.2
Melting Point 884°C
Quantity 12kg
Physical Form Solid
Viscosity [NULL]
Purity Grade USP
Molecular Formula Na2O4S · 10 H2O
Color White
pH 5.2 to 9.2
Density 1.46g/cm³
Heavy Metals (as Pb) 0.001% max.
Chemical Name or Material Sodium Sulfate Decahydrate
ChemAlert Storage Symbol Gray
Identification Pass Test
Packaging Poly Pail
MDL Number MFCD00149189
Acidity Alkalinity Pass Test
Loss on Drying 51.0 to 57.0%
Assay Percent Range ≥99.0 %
CAS 7727-73-3
Chloride 0.02% max.

Specifications

Chemical Name or Material Sodium Sulfate Decahydrate
CAS 7727-73-3
Purity Grade USP
Physical Form Solid
Assay ≥99.0%
Assay Percent Range ≥99.0 %
Molecular Formula Na2O4S · 10 H2O
Formula Weight 322.2
MDL Number MFCD00149189
Accessories
 

SDS & Safety Documents