Please login to your online account to display your discounted pricing

Sodium Sulfate Decahydrate (Granular/USP), Fisher Chemical

$2,063.64 - $2,063.64

Chemical Identifiers

Chemical Name or Material Sodium Sulfate Decahydrate
CAS 7727-73-3
Purity Grade USP
Physical Form Solid
Assay ≥99.0%
Assay Percent Range ≥99.0 %
Molecular Formula Na2O4S · 10 H2O
Formula Weight 322.2
MDL Number MFCD00149189
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S414-12

  SDS
fisher scientific company
FLS41412
Poly Pail 12kg Each for $2,063.64
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 322.2
Melting Point 884°C
Quantity 12kg
Physical Form Solid
Density 1.46g/cm³
Molecular Formula Na2O4S · 10 H2O
Purity Grade USP
Color White
pH 5.2 to 9.2
Heavy Metals (as Pb) 0.001% max.
Chemical Name or Material Sodium Sulfate Decahydrate
ChemAlert Storage Symbol Gray
MDL Number MFCD00149189
Packaging Poly Pail
Identification Pass Test
Acidity Alkalinity Pass Test
Chloride 0.02% max.
Loss on Drying 51.0 to 57.0%
Assay Percent Range ≥99.0 %
CAS 7727-73-3