Please login to your online account to display your discounted pricing

Sodium Sulfate Decahydrate (Granular/USP), Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLS414 500

 View more versions of this product

Catalog No. S414-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Acidity Alkalinity Pass Test
Chloride 0.02% max.
Density 1.46g/cm³
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Loss on Drying 51.0 to 57.0%
Packaging Poly Bottle
Viscosity [NULL]
Color White
Melting Point 884°C / 1623.2°F
pH 5.2 to 9.2
Quantity 500g
Chemical Name or Material Sodium Sulfate Decahydrate
CAS 7727-73-3
Purity Grade USP
Physical Form Solid
Assay 99.0% min.
Assay Percent Range ≥99.0 %
Molecular Formula Na2O4S · 10 H2O
Formula Weight 322.2g/mol
MDL Number MFCD00149189
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Sodium Sulfate Decahydrate
CAS 7727-73-3
Purity Grade USP
Physical Form Solid
Assay 99.0% min.
Assay Percent Range ≥99.0 %
Molecular Formula Na2O4S · 10 H2O
Formula Weight 322.2g/mol
MDL Number MFCD00149189
Accessories
 

SDS & Safety Documents