Please login to your online account to display your discounted pricing

Sodium Sulfate, Decahydrate, Certified, 22.2% (w/v), LabChem™

$75.89 - $75.89

Chemical Identifiers

Chemical Name or Material Sodium Sulfate
CAS 7732-18-5
CAS Max % 90.2
CAS 7757-82-6
CAS Max % 9.8
Name Note Decahydrate
Purity Grade Certified Grade
Linear Formula Na2SO4
Formula Weight 142.04
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC249002

 
lab chem
LC249002
1L Each for $75.89
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 142.04
CAS Max % 9.8
Assay 22.2% (w/v)
Quantity 1L
Solubility Soluble in water
Identification Passes Test
Decomposition Information Sulfur compounds
Storage Information Room Temperature
Color Clear, colorless
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Chemical Name or Material Sodium Sulfate
CAS 7732-18-5
Linear Formula Na2SO4
Name Note Decahydrate
CAS Max % 90.2
CAS 7757-82-6
Physical Form Liquid
Packaging Poly Bottle
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.