missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium sulfate decahydrate, 99+%, for analysis, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC451100050

 View more versions of this product

Catalog No. AC451100050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sodium sulfate decahydrate
99+%
Na2SO4·10H2O
Solubility in water: soluble
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]
322.2
CHEBI:32586
99+%
5ppm max
10ppm max
5kg
7727-73-3
Na2O410H2O
disodium sulfate decahydrate, disodium sulfate decahydrate na2so4.10h2o, glauber's salt, sodium sulfate decahydrate, sodium sulfate decahydrate na2so4.10h2o, sodium sulfate na2so4 decahydrate, sodium sulfate usp, sodium sulphate decahydrate, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j
RSIJVJUOQBWMIM-UHFFFAOYSA-L
disodium;sulfate;decahydrate
62649
322.2
Analysis
Plastic bottle
884.0°C

Chemical Identifiers

7727-73-3
322.2
disodium sulfate decahydrate, disodium sulfate decahydrate na2so4.10h2o, glauber's salt, sodium sulfate decahydrate, sodium sulfate decahydrate na2so4.10h2o, sodium sulfate na2so4 decahydrate, sodium sulfate usp, sodium sulphate decahydrate, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j
CHEBI:32586
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]
Na2O410H2O
RSIJVJUOQBWMIM-UHFFFAOYSA-L
62649
disodium;sulfate;decahydrate
SDS Documents