missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium sulfate decahydrate, 99+%, for analysis, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451100010
Specifications
Sodium sulfate decahydrate | |
99+% | |
Na2SO4·10H2O | |
Solubility in water: soluble | |
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] | |
322.2 | |
CHEBI:32586 | |
99+% | |
5ppm max | |
10ppm max | |
1kg |
7727-73-3 | |
Na2O4S·10H2O | |
disodium sulfate decahydrate, disodium sulfate decahydrate na2so4.10h2o, glauber's salt, sodium sulfate decahydrate, sodium sulfate decahydrate na2so4.10h2o, sodium sulfate na2so4 decahydrate, sodium sulfate usp, sodium sulphate decahydrate, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j | |
RSIJVJUOQBWMIM-UHFFFAOYSA-L | |
disodium;sulfate;decahydrate | |
62649 | |
322.2 | |
Analysis | |
Plastic bottle | |
884.0°C |
Chemical Identifiers
7727-73-3 | |
322.2 | |
disodium sulfate decahydrate, disodium sulfate decahydrate na2so4.10h2o, glauber's salt, sodium sulfate decahydrate, sodium sulfate decahydrate na2so4.10h2o, sodium sulfate na2so4 decahydrate, sodium sulfate usp, sodium sulphate decahydrate, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j | |
CHEBI:32586 | |
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Na2O4S·10H2O | |
RSIJVJUOQBWMIM-UHFFFAOYSA-L | |
62649 | |
disodium;sulfate;decahydrate |