missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Sodium Sulfate Anhydrous, Certified ACS, Suitable for Pesticide Residue Analysis (Granular, 10-60 mesh), Fisher Chemical™
Manufacturer: Fisher Chemical S415200LB
Specifications
Sodium Sulfate Anhydrous | |
White | |
5 to 8 | |
Suitable for pesticide residue analysis | |
Na2O4S | |
disodium sulfate, disodium sulphate, salt cake, sodium sulfate, sodium sulfate 2:1, sodium sulfate anhydrous, sodium sulfate, anhydrous, sodium sulphate, sulfuric acid disodium salt, thenardite | |
[O-]S(=O)(=O)[O-].[Na+].[Na+] | |
142.036 | |
CHEBI:32149 | |
Solid | |
Certified ACS | |
Pass Test | |
0.001% max. | |
0.005% max. | |
5ppm max. | |
0.001% max. | |
Pass Test | |
0.001% max. |
7757-82-6 | |
884°C | |
200 lb. | |
≥99.0 % | |
MFCD00003504 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium;sulfate | |
24436 | |
142.04 | |
≥99.0% | |
5ppm max. | |
0.01% max. | |
0.5% max. | |
10 to 60 Mesh | |
Fiber Drum | |
0.01% max. | |
0.01% max. | |
Decomposition Information: >890°C |
Chemical Identifiers
7757-82-6 | |
142.036 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
24436 | |
disodium;sulfate |
Na2O4S | |
MFCD00003504 | |
disodium sulfate, disodium sulphate, salt cake, sodium sulfate, sodium sulfate 2:1, sodium sulfate anhydrous, sodium sulfate, anhydrous, sodium sulphate, sulfuric acid disodium salt, thenardite | |
CHEBI:32149 | |
[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Safety and Handling

CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0
SIGNAL WORD: CAUTION!