Please login to your online account to display your discounted pricing

Sodium Sulfate Anhydrous ACS Reagent Grade, Ricca Chemical

$81.97 - $108.91

Chemical Identifiers

Chemical Name or Material Sodium Sulfate
Name Note Anhydrous
CAS 7757-82-6
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Physical Form Crystals
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RDCS0850500

 
ricca chemical company
RIRDCS0850500B1
Poly wht WM 500g Each for $81.97

RDCS0850100

 
ricca chemical company
RIRDCS08501000B1
Poly wht WM 1000g Each for $108.91
Description & Specifications

Specifications

Quantity 500g
Physical Form Crystals
Purity Grade ACS Reagent Grade
Color White
CAS Min % 98.0
CAS Max % 100.0
CAS 7757-82-6
Packaging Poly wht WM
Name Note Anhydrous
Chemical Name or Material Sodium Sulfate
Shelf Life 60 months
Safety and Handling

60 months