missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium sulfate, anhydrous, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 8761106*

 View more versions of this product

Catalog No. AA8761106



Specifications

Specifications

Sodium sulfate
7757-82-6
(metals basis)
Na2SO4
14,8680
[O-]S(=O)(=O)[O-].[Na+].[Na+]
142.036
CHEBI:32149
Powder/Chunks
Odorless
Hygroscopic
884°C
Anhydrous
99.99%
Na2O4S
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
142.04
2.680g/mL
1.4840
White
5g

Chemical Identifiers

7757-82-6
142.036
PMZURENOXWZQFD-UHFFFAOYSA-L
CHEBI:32149
[O-]S(=O)(=O)[O-].[Na+].[Na+]
Na2O4S
MFCD00003504
24436
disodium;sulfate
Safety and Handling

Safety and Handling

RTECSNumber : WE1650000

TSCA : TSCA

SDS Documents