Please login to your online account to display your discounted pricing

Sodium Sulfate, Anhydrous, 99.0% min., ACS, EMD Millipore


Quantity 50kg

Manufacturer: emd millipore corporation  SX0760F20

 View more versions of this product

Catalog No. MSX0760F20

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Granules
Formula Weight 142.04
Color White
Quantity 50kg
Molecular Formula Na2SO4
CAS 7757-82-6
Chemical Name or Material Sodium Sulfate, Anhydrous
Name Note For ACS analysis
Assay Percent Range 99.0% min.

Specifications

Chemical Name or Material Sodium Sulfate, Anhydrous
Name Note For ACS analysis
CAS 7757-82-6
Physical Form Granules
Assay Percent Range 99.0% min.
Molecular Formula Na2SO4
Formula Weight 142.04