Please login to your online account to display your discounted pricing

Sodium Sulfate, Anhydrous, 99.0% min., ACS, EMD Millipore

$207.43 - $1,810.49

Chemical Identifiers

Chemical Name or Material Sodium Sulfate, Anhydrous
Name Note For ACS analysis
CAS 7757-82-6
Physical Form Granules
Assay Percent Range 99.0% min.
Molecular Formula Na2SO4
Formula Weight 142.04
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MSX0760E5

 
emd millipore corporation
SX0760E5
12kg Each for $737.29

MSX07603

 
emd millipore corporation
SX07603
2.5kg Each for $207.43

MSX0760F20

 
emd millipore corporation
SX0760F20
50kg Each for $1,810.49
Description & Specifications

Specifications

Formula Weight 142.04
Physical Form Granules
Color White
Quantity 12kg
Name Note For ACS analysis
Molecular Formula Na2SO4
Chemical Name or Material Sodium Sulfate, Anhydrous
Assay Percent Range 99.0% min.
CAS 7757-82-6