Please login to your online account to display your discounted pricing

Sodium Sulfate, Anhydrous, 98.0% min., EMD Millipore


Quantity 2.5kg

Manufacturer: emd millipore corporation  SX07633

 View more versions of this product

Catalog No. MSX07633

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS/NF/USP
Formula Weight 142.4
Quantity 2.5kg
Color White
Molecular Formula Na2SO4
CAS 7757-82-6
Assay Percent Range 98.0%, min.
Linear Formula Na2SO5
Chemical Name or Material Sodium Sulfate, Anhydrous
Physical Form Powder

Specifications

Chemical Name or Material Sodium Sulfate, Anhydrous
CAS 7757-82-6
Purity Grade ACS/NF/USP
Physical Form Powder
Assay Percent Range 98.0%, min.
Molecular Formula Na2SO4
Linear Formula Na2SO5
Formula Weight 142.4