missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Sulfate, Anhydrous, 98.0% min., EMD Millipore

$90.83 - $176.89

Chemical Identifiers

CAS 7757-82-6
Physical Form Powder
Assay Percent Range 98.0%, min.
Molecular Formula Na2SO4
Linear Formula Na2SO5
Formula Weight 142.4
Chemical Name or Material Sodium Sulfate, Anhydrous
Grade ACS/NF/USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MSX07631 Emd Millipore Corporation
SX07631
500g Each for $90.83
MSX07633 Emd Millipore Corporation
SX07633
2.5kg Each for $176.89
Specifications Description & Specifications

Chemical Identifiers

7757-82-6
98.0%, min.
Na2SO5
Sodium Sulfate, Anhydrous
Powder
Na2SO4
142.4
ACS/NF/USP

Specifications

7757-82-6
98.0%, min.
Na2SO5
White
Sodium Sulfate, Anhydrous
Powder
Na2SO4
142.4
500g
ACS/NF/USP