Please login to your online account to display your discounted pricing

Sodium Sulfate Anhydrous

$11.35 - $11.35

Chemical Identifiers

Chemical Name or Material Sodium Sulfate
CAS 7757-82-6
Linear Formula Na2SO4
Formula Weight 142.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S75245

 
fisher science education
SS0830-100G
Lab Grade - Powder 100g Each for $11.35
Description & Specifications

Specifications

Formula Weight 142.04
Quantity 100g
Color Blue
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Anhydrous or Hydrate Anhydrous
Shelf Life Indefinite
CAS 7757-82-6
ChemAlert Storage Symbol Gray
Chemical Name or Material Sodium Sulfate
Packaging Lab Grade - Powder
Linear Formula Na2SO4
Safety and Handling
 

Indefinite