missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Sodium Sulfate Anhydrous
Manufacturer: Fisher Science Education SS0830100G
Specifications
Sodium Sulfate | |
Blue | |
Na2O4S | |
disodium sulfate, disodium sulphate, salt cake, sodium sulfate, sodium sulfate 2:1, sodium sulfate anhydrous, sodium sulfate, anhydrous, sodium sulphate, sulfuric acid disodium salt, thenardite | |
[O-]S(=O)(=O)[O-].[Na+].[Na+] | |
142.036 | |
CHEBI:32149 | |
Lab Grade - Powder | |
Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment. |
7757-82-6 | |
100g | |
Na2SO4 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium;sulfate | |
24436 | |
142.04 | |
Anhydrous |
Safety and Handling

ShelfLife : Indefinite