missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium sulfate, ACS, 99.0% min. , Alfa Aesar™

For Use in Drying organic Liquids, in Kjeldahl Nitrogen Determination, in Manufacturing of Glass

Manufacturer:  Alfa Aesar 11560A9

 View more versions of this product

Catalog No. AA11560A9



Specifications

Specifications

Sodium sulfate
7757-82-6
Na2O4S
White powder
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
142.04
ACS
White
For ACS analysis
99.0% min.
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
[O-]S(=O)(=O)[O-].[Na+].[Na+]
142.036
CHEBI:32149
Granular
Odorless
10kg

Chemical Identifiers

7757-82-6
142.036
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
disodium;sulfate
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149
[O-]S(=O)(=O)[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

EINECSNumber : 231-820-9

Documents