Please login to your online account to display your discounted pricing

Sodium sulfate, ACS, 99.0% min, Alfa Aesar™

In drying organic liquids, in Kjeldahl nitrogen determination, in manufacturing of glass

$57.27 - $57.27

Chemical Identifiers

Chemical Name or Material Sodium sulfate
CAS 7757-82-6
Purity Grade ACS Grade
Molecular Formula Na2SO4
Formula Weight 142.04
MDL Number MFCD00003504
Merck Index 14,8680
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1156036

 
alfa aesar
1156036
Each for $57.27
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 142.04
Quantity 500g
Chemical Name or Material Sodium sulfate
TSCA TSCA
Odor Odorless
Physical Form Granular
Refractive Index 1.484
Assay ≥99.0%
Sensitivity Hygroscopic
Molecular Formula Na2SO4
MDL Number MFCD00003504
CAS 7757-82-6
Merck Index 14,8680
Melting Point 884°C
Solubility Information Soluble in water. Insoluble in alcohol
Density 2.680
RTECS Number WE1650000