missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium sulfate, 99%, extra pure, anhydrous, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 196640250

 View more versions of this product

Catalog No. AC196640250



Description

Description

Specifications

Specifications

Sodium sulfate
98.5% min
Na2SO4
01,1103
disodium sulfate, disodium sulphate, salt cake, sodium sulfate, sodium sulfate 2:1, sodium sulfate anhydrous, sodium sulfate, anhydrous, sodium sulphate, sulfuric acid disodium salt, thenardite
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
142.04
99%
2.68
Plastic bottle
Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
0.1% max.
White
25g
7757-82-6
Na2O4S
MFCD00003504
15, 8809
Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide,insoluble in alcohol
[O-]S(=O)(=O)[O-].[Na+].[Na+]
142.04
CHEBI:32149
Crystalline Powder
Extra Pure
10ppm max.
3000ppm max.
200ppm max.
>1700.0°C
884.0°C

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
disodium;sulfate
Na2O4S
MFCD00003504
disodium sulfate, disodium sulphate, salt cake, sodium sulfate, sodium sulfate 2:1, sodium sulfate anhydrous, sodium sulfate, anhydrous, sodium sulphate, sulfuric acid disodium salt, thenardite
CHEBI:32149
[O-]S(=O)(=O)[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

EINECSNumber : 231-820-9

SDS Documents