missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium sulfate, +99%, for analysis ACS, anhydrous, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC424410100

 View more versions of this product

Catalog No. AC424410100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Sodium sulfate
ACS Reagent Grade
≥99.0%
Na2O4S
142.04
15, 8809
10ppm max.
0.01% max.
0.5% max.
5ppm max.
10ppm max.
Decomposition Information: >890°C
5.2 to 9.2 (5% soln. at 25°C)
7757-82-6
Granular Crystalline Powder
99+%
Na2SO4
01,1103
100ppm max.
5ppm max.
10ppm max.
50ppm max.
Plastic Bucket
100ppm max.
White
10kg

Chemical Identifiers

7757-82-6
Na2O4S
142.04
≥99.0%
99+%
Na2SO4
Sodium sulfate
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0

SIGNAL WORD: CAUTION!