missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium sulfate, 99%, for biochemistry, anhydrous, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC447925000
Specifications
Sodium sulfate | |
99% | |
Na2SO4 | |
disodium sulfate, disodium sulphate, salt cake, sodium sulfate, sodium sulfate 2:1, sodium sulfate anhydrous, sodium sulfate, anhydrous, sodium sulphate, sulfuric acid disodium salt, thenardite | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium;sulfate | |
24436 | |
142.04 | |
Biochemistry | |
0.01% max | |
0.001% max | |
>1700.0°C | |
500g |
7757-82-6 | |
Na2O4S | |
15,8809 | |
Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol | |
[O-]S(=O)(=O)[O-].[Na+].[Na+] | |
142.04 | |
CHEBI:32149 | |
99% | |
0.001% max | |
Plastic bottle | |
(10% in water) Clear colorless | |
884.0°C |
Chemical Identifiers
7757-82-6 | |
142.04 | |
disodium sulfate, disodium sulphate, salt cake, sodium sulfate, sodium sulfate 2:1, sodium sulfate anhydrous, sodium sulfate, anhydrous, sodium sulphate, sulfuric acid disodium salt, thenardite | |
CHEBI:32149 | |
[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Na2O4S | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
24436 | |
disodium;sulfate |
Safety and Handling
EINECSNumber : 231-820-9