Please login to your online account to display your discounted pricing

Sodium Sulfate, 22.2% (w/v) Aqueous Solution, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR780000020F

 View more versions of this product

Catalog No. R780000020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear colorless
Physical Form Liquid
Quantity 20L
Chemical Name or Material Sodium Sulfate
CAS 7732-18-5
CAS Min % 80.0
CAS Max % 83.0
CAS 7757-82-6
Packaging Cubitainer
CAS Min % 17.81
CAS Max % 18.54

Specifications

Chemical Name or Material Sodium Sulfate
CAS 7732-18-5
CAS Min % 80.0
CAS Max % 83.0
CAS 7757-82-6
CAS Min % 17.81
CAS Max % 18.54
Physical Form Liquid