Please login to your online account to display your discounted pricing

Sodium sulfanilate hydrate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Sodium sulfanilate hydrate
Name Note 97%
CAS 123333-70-0
Assay ≥96.0% (on dry substance)
Assay Percent Range 97%
Molecular Formula C6H6NNaO3S·xH2O
Formula Weight 195.17
MDL Number MFCD00151043
Synonym Sulfanilic acid sodium salt
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC346882500

  SDS
fisher scientific acros
346882500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0% (on dry substance)
Formula Weight 195.17
Physical Form Crystalline Powder
Quantity 250g
Color White to pink or light brown
Water 16% max. (K.F.)
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym Sulfanilic acid sodium salt
Name Note 97%
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6H6NNaO3S·xH2O
Chemical Name or Material Sodium sulfanilate hydrate
MDL Number MFCD00151043
CAS 123333-70-0
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.