Learn More
Sodium succinate hexahydrate, 99%, Thermo Scientific Chemicals
$86.14 - $801.08
Chemical Identifiers
CAS | 6106-21-4 |
---|---|
Molecular Formula | C4H16Na2O10 |
Molecular Weight (g/mol) | 270.142 |
MDL Number | MFCD00149117 |
InChI Key | ZBTUYCUNQBRXOR-UHFFFAOYSA-L |
Synonym | disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate |
PubChem CID | 3083938 |
ChEBI | CHEBI:63686 |
IUPAC Name | disodium;butanedioate;hexahydrate |
SMILES | C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4198330
|
Thermo Scientific Chemicals
04198330 |
250 g |
Each for $86.14
|
|
|||||
AA41983A1
|
Thermo Scientific Chemicals
041983A1 |
1 kg |
Each for $228.48
|
|
|||||
AA41983A7
|
Thermo Scientific Chemicals
041983A7 |
5 kg |
Each for $801.08
|
|
|||||
Description
Sodium succinate hexahydrate is used as a tricarboxylic acid cycle (Krebs cycle) supplement in cell culture applications. It is also used as a flavoring agent for food and beverages. It serves as an intermediate for dyes, perfumes, lacquers, photographic chemicals, alkyd resins, plasticizer, metal treatment chemical, vehicle water cooling systems and coatings. Further, it is used to study x-ray crystallography, protein structural analysis, proteomics and optimization reagents. It is utilized to determine the calorimetric and diffractometric evidence for the sequential crystallization of buffer components.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6106-21-4 | |
270.142 | |
ZBTUYCUNQBRXOR-UHFFFAOYSA-L | |
3083938 | |
disodium;butanedioate;hexahydrate |
C4H16Na2O10 | |
MFCD00149117 | |
disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate | |
CHEBI:63686 | |
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+] |
Specifications
6106-21-4 | |
C4H16Na2O10 | |
MFCD00149117 | |
3921044 | |
14,8679 | |
Soluble in water. Insoluble in alcohol. | |
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+] | |
270.142 | |
CHEBI:63686 | |
99% | |
Sodium Succinate Hexahydrate |
120°C to 6H2O | |
NaO2CCH2CH2CO2Na·6H2O | |
250 g | |
Hygroscopic | |
disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate | |
ZBTUYCUNQBRXOR-UHFFFAOYSA-L | |
disodium;butanedioate;hexahydrate | |
3083938 | |
270.14 (162.05 Anhydrous) | |
Crystalline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-778-7
RTECSNumber : WM7751000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only