missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium succinate hexahydrate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 033386A1

 View more versions of this product

Catalog No. AA33386A1



Description

Description

Sodium succinate hexahydrate is used as an ingredient of topical preparations for the treatment of cataract.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium succinate hexahydrate
120°C (in -6H2O)
C4H16Na2O10
MFCD00149117
3921044
14,8679
It is soluble in water at a concentration of 270g/L at 20°C (68°F) .
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]
270.142
CHEBI:63686
Crystalline or Granular Powder
6106-21-4
White
NaO2CCH2CH2CO2Na·6H2O
1 kg
Hygroscopic
disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
disodium;butanedioate;hexahydrate
3083938
270.14 (162.05 Anhydrous)

Chemical Identifiers

6106-21-4
270.142
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
3083938
disodium;butanedioate;hexahydrate
C4H16Na2O10
MFCD00149117
disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
CHEBI:63686
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 205-778-7

RTECSNumber : WM7751000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only