missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Standard, 1000mg/L in 2% HNO3, (Assurance Grade), SPEX CertiPrep

Supplier:  SPEX CertiPrep Group LLC PLNA22T

 View more versions of this product

Catalog No. PLNA22T



Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Sodium Standard
  • Made with ASTM Type I Water
  • 1,000 μg/mL concentration
  • 250 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Specifications

Specifications

Sodium Standard
Poly Bottle
Na
KEAYESYHFKHZAL-UHFFFAOYSA-N
sodium
5360545
Assurance
7440-23-5,7732-18-5,7697-37-2
250 mL
MFCD00085307
[Na]
22.99
CHEBI:26708

Chemical Identifiers

7440-23-5
22.99
KEAYESYHFKHZAL-UHFFFAOYSA-N
CHEBI:26708
[Na]
Na
MFCD00085307
5360545
sodium
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions