Please login to your online account to display your discounted pricing

Sodium Standard, 10,000μg/mL in water with dilute nitric acid, Ultra Scientific

$182.54 - $182.54

Chemical Identifiers

Chemical Name or Material Sodium Standard
CAS 007732-18-5
CAS Max % 97.0
CAS 007697-37-2
CAS Max % 2.0
CAS 007440-23-5
CAS Max % 1.0
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-ICP-111

 
ultra scientific
ICP111
125mL Each for $182.54
Description & Specifications

Specifications

Quantity 125mL
Analyte or Component Names Sodium,water,nitric acid
Solvent or Matrix Water with dilute nitric acid
CAS Max % 97.0
Concentration or Composition (by Analyte or Components) 10, 000μg/mL
Chemical Name or Material Sodium Standard
CAS 007697-37-2
CAS Max % 2.0
CAS 007440-23-5
CAS Max % 1.0
CAS 007732-18-5