Please login to your online account to display your discounted pricing

Sodium-silica gel, 35-40% alkali metal in silica gel, Stage II, Alfa Aesar™

Reductively Produces Radical Anions and Dianions, Dechlorinates Alkyl and Aryl Halides and Desulfurize Compounds. Also Ideal For Reduction of Water To Produce Hydrogen Gas, Solvent Drying and Anti-Oxidant

Chemical Identifiers

Chemical Name or Material Sodium-silica gel
Name Note 35-40% alkali metal in silica gel, Stage II
Physical Form Powder
MDL Number MFCD01941718
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4489414

  SDS
alfa aesar
4489414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4489406

  SDS
alfa aesar
4489406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4489422

  SDS
alfa aesar
4489422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 2 GHS H Statement
H228-H261-H314-H318
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P231+P232-P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P405-P501a
Physical Form Powder
Quantity 25g
Color Black
DOT Information DOT Class: 4.3, Packing Group: II
Density 0.7g/cm3
Chemical Name or Material Sodium-silica gel
Name Note 35-40% alkali metal in silica gel, Stage II
Sensitivity Moisture sensitive
MDL Number MFCD01941718
Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P405-P501a

GHS H Statement
H228-H261-H314-H318