Please login to your online account to display your discounted pricing

Sodium-silica gel, 35-40% alkali metal in silica gel, Stage I, Alfa Aesar™

Reductively Produces Radical Anions and Dianions, Dechlorinates Alkyl and Aryl Halides and Desulfurize Compounds

Chemical Identifiers

Chemical Name or Material Sodium-silica gel
Name Note 35-40% alkali metal in silica gel, Stage I
Physical Form Powder
MDL Number MFCD01941718
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4489314

 
alfa aesar
4489314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4489322

 
alfa aesar
4489322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Quantity 25g
Color Black
DOT Information DOT Class: 4.3, Packing Group: II
Chemical Name or Material Sodium-silica gel
Name Note 35-40% alkali metal in silica gel, Stage I
Sensitivity Moisture sensitive
MDL Number MFCD01941718
Health Hazard 1 H228-H261-H314-H318: Flammable solid. In contact with water releases flammable gas. Causes severe skin burns and eye damage. Causes serious eye damage.
Health Hazard 2 Keep container tightly closed and dry. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Take precautionary measures against static discharges. Wear suitable protective clothing
Density 0.7g/cm3
Safety and Handling

H228-H261-H314-H318: Flammable solid. In contact with water releases flammable gas. Causes severe skin burns and eye damage. Causes serious eye damage.

Keep container tightly closed and dry. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Take precautionary measures against static discharges. Wear suitable protective clothing