Learn More
Sodium selenite, 44-46% Se, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 200730250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium selenite | |
Anhydrous | |
White to Beige | |
25 g | |
Na2SeO3 | |
15, 8802 | |
Solubility in water: 950g/L (20°C). Other solubilities: insoluble in alcohol | |
[O-][Se](=O)[O-].[Na+].[Na+] | |
172.94 | |
CHEBI:48843 | |
44 to 46% Se | |
Glass bottle |
350°C | |
10102-18-8 | |
Powder | |
Na2O3Se | |
MFCD00003489 | |
sodium selenite, disodium selenite, natriumselenit, selenious acid, disodium salt, natriumselenit german, disodium selenium trioxide, unii-hiw548rq3w, selenious acid disodium salt, ccris 1260, hsdb 768 | |
BVTBRVFYZUCAKH-UHFFFAOYSA-L | |
disodium;selenite | |
24934 | |
172.94 | |
0.5% max. |
Chemical Identifiers
10102-18-8 | |
172.94 | |
BVTBRVFYZUCAKH-UHFFFAOYSA-L | |
24934 | |
disodium;selenite |
Na2O3Se | |
MFCD00003489 | |
sodium selenite, disodium selenite, natriumselenit, selenious acid, disodium salt, natriumselenit german, disodium selenium trioxide, unii-hiw548rq3w, selenious acid disodium salt, ccris 1260, hsdb 768 | |
CHEBI:48843 | |
[O-][Se](=O)[O-].[Na+].[Na+] |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Contact with acids liberates toxic gas.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear eye protection/face protection.
Do not breathe
GHS Signal Word: Danger
EINECSNumber : 233-267-9
RUO – Research Use Only