missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium selenate, anhydrous, >99.8% (metals basis), Alfa Aesar™

For Use As a Reagent

Manufacturer:  Alfa Aesar 12613A1

 View more versions of this product

Catalog No. AA12613A1



Specifications

Specifications

Sodium selenate, anhydrous
99.8+% (metals basis)
MFCD00003490
sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
[O-][Se](=O)(=O)[O-].[Na+].[Na+]
188.947
CHEBI:77775
Powder
Purple
13410-01-0
Na2O4Se
White powder
MHQOTKLEMKRJIR-UHFFFAOYSA-L
disodium;selenate
25960
188.94
Odorless
1kg

Chemical Identifiers

13410-01-0
188.947
MHQOTKLEMKRJIR-UHFFFAOYSA-L
25960
disodium;selenate
Na2O4Se
MFCD00003490
sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
CHEBI:77775
[O-][Se](=O)(=O)[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

H300-H330-H373-H400-H410: Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: I

EINECSNumber : 236-501-8

Documents