Please login to your online account to display your discounted pricing

Sodium Rod, Diameter 2.5cm (Protective Liquid: Paraffin Oil), EMD Millipore™


Manufacturer: emd millipore corporation  1.06260.1000

 View more versions of this product

Catalog No. M1062601000

  • / Each
 

Description & Specifications

Chemical Identifiers

Appearance Silver solid
Odor Odorless
Packaging Glass bottle
Chemical Name or Material Sodium
CAS 7440-23-5
Physical Form Solid
Molecular Formula Na (Hill)
Formula Weight 22.99g/mol. (molar mass)
UN Number UN 1428
Health Hazard 1 H260: In contact with water releases flammable gases which may ignite spontaneously.
H314: Causes severe skin burns and eye damage.
EUH014: Reacts violently with water.
DOT Information DOT Class: 4.3, Packaging Group: I
EINECS Number 231-132-9
Accessories

H260: In contact with water releases flammable gases which may ignite spontaneously.
H314: Causes severe skin burns and eye damage.
EUH014: Reacts violently with water.