missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Rod, Diameter 2.5cm (Protective Liquid: Paraffin Oil), EMD Millipore™

$509.60 - $509.60

Chemical Identifiers

CAS 7440-23-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1062600250 View Documents Millipore Sigma
1062600250
250g Glass bottle Each for $509.60
Specifications

Chemical Identifiers

7440-23-5
22.99
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
CHEBI:26708
[Na]
Na
KEAYESYHFKHZAL-UHFFFAOYSA-N
5360545
sodium

Specifications

250g
Silver solid
UN 1428
KEAYESYHFKHZAL-UHFFFAOYSA-N
[Na]
Glass bottle
5360545
22.99. (molar mass)
Sodium
7440-23-5
Na
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
Odorless
sodium
22.99
CHEBI:26708
Solid
Safety and Handling

Safety and Handling

H260: In contact with water releases flammable gases which may ignite spontaneously.
H314: Causes severe skin burns and eye damage.
EUH014: Reacts violently with water.

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 231-132-9