Please login to your online account to display your discounted pricing

Sodium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


For atomic absorption. (1mL = 1mg Na) Solute: Sodium Chloride Solvent: Distilled Water

Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SS139500

 View more versions of this product

Catalog No. SS139-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Boiling Point 100°C
Melting Point 0°C
Quantity 500mL
Color Colorless
Physical Form Liquid
Density 1.0g/cm³
Vapor Pressure 14mmHg
CAS 7732-18-5
MDL Number MFCD00085307
Packaging Poly Bottle
Molecular Formula NaCl
Traceability to NIST LOT NUMBER USED SODIUM REF. STD.
Concentration 990 to 1010ppm
ChemAlert Storage Symbol Gray
Name Note 1000ppm ±1%
CAS 7647-14-5
Chemical Name or Material Sodium, Reference Standard Solution

Specifications

Chemical Name or Material Sodium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7647-14-5
CAS 7732-18-5
Purity Grade Certified Grade
Molecular Formula NaCl
MDL Number MFCD00085307
Accessories
 

SDS & Safety Documents