missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium pyruvate, 99%

Manufacturer:  Alfa Aesar™ A1114830

 View more versions of this product

Catalog No. AAA1114830


 

Description

Description

Applications
Sodium pyruvate is used in cell culture media as an additional source of energy. It acts as an antioxidant and finds protective effects against oxygen radicals. It serves as an intermediate in many metabolic pathways such as sugar metabolism. It is converted into acetyl coenzyme A and enters the trichloroacetic acid cycle. In addition, it is involved with amino acid metabolism and initiates the Kreb′s cycle. It plays an important role as a free radical scavenger.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Sodium pyruvate
99%
MFCD00002586
Pyruvic acid sodium salt
CC(=O)C(=O)[O-].[Na+]
110.044
CHEBI:50144
>300°C
113-24-6
C3H3NaO3
3568341
DAEPDZWVDSPTHF-UHFFFAOYSA-M
sodium;2-oxopropanoate
23662274
110.04
250g

Chemical Identifiers

113-24-6
110.044
DAEPDZWVDSPTHF-UHFFFAOYSA-M
23662274
C3H3NaO3
MFCD00002586
Pyruvic acid sodium salt
CHEBI:50144
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents