missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium pyrophosphate decahydrate, 99-103%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 448065000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
93.8°C | |
80.0°C | |
500 g | |
Na4O7P2·10H2O | |
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
tetrasodium;phosphonato phosphate;decahydrate | |
3084150 | |
99 to 103% | |
Plastic bottle |
Sodium pyrophosphate decahydrate | |
13472-36-1 | |
99-103% | |
Na4P2O7·10H2O | |
Solubility in water: 62g/L (20°C). Other solubilities: insoluble in ammonia, insoluble in alcohol | |
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] | |
446.05 | |
446.05 | |
ACS Reagent |
Chemical Identifiers
13472-36-1 | |
446.05 | |
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
tetrasodium;phosphonato phosphate;decahydrate |
Na4O7P2·10H2O | |
VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
3084150 | |
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only