Please login to your online account to display your discounted pricing

Sodium Pyrophosphate, Decahydrate, 99.0% min., ACS, EMD Millipore


Quantity 12kg

Manufacturer: emd millipore corporation  SX07405

 View more versions of this product

Catalog No. MSX07405

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 446.6
Purity Grade ACS/EP/Reagent Grade
Quantity 12kg
Bulk Density 650kg/m3
Density 1.82g/cm3 (20°C)
Synonym TSPP
CAS 13472-36-1
Name Note EMSURE™, For Analysis
Linear Formula Na4P2O7 · 10H2O
Recommended Storage Store without restrictions
Assay Percent Range 99.0% min.
Chemical Name or Material Sodium Pyrophosphate, Decahydrate
Molecular Formula Na4O7P2 · 10H2O

Specifications

Chemical Name or Material Sodium Pyrophosphate, Decahydrate
Name Note EMSURE™, For Analysis
CAS 13472-36-1
Purity Grade ACS/EP/Reagent Grade
Assay Percent Range 99.0% min.
Molecular Formula Na4O7P2 · 10H2O
Linear Formula Na4P2O7 · 10H2O
Formula Weight 446.6
Synonym TSPP
Accessories

Store without restrictions