Please login to your online account to display your discounted pricing

Sodium Pyrophosphate, Decahydrate, 99.0% min., ACS, EMD Millipore

$1,827.66 - $1,827.66

Chemical Identifiers

Chemical Name or Material Sodium Pyrophosphate, Decahydrate
Name Note EMSURE™, For Analysis
CAS 13472-36-1
Purity Grade ACS/EP/Reagent Grade
Assay Percent Range 99.0% min.
Molecular Formula Na4O7P2 · 10H2O
Linear Formula Na4P2O7 · 10H2O
Formula Weight 446.6
Synonym TSPP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MSX07405

 
emd millipore corporation
SX07405
12kg Each for $1,827.66
Description & Specifications

Specifications

Purity Grade ACS/EP/Reagent Grade
Formula Weight 446.6
Quantity 12kg
Bulk Density 650kg/m3
Density 1.82g/cm3 (20°C)
Chemical Name or Material Sodium Pyrophosphate, Decahydrate
Name Note EMSURE™, For Analysis
Assay Percent Range 99.0% min.
Molecular Formula Na4O7P2 · 10H2O
Linear Formula Na4P2O7 · 10H2O
CAS 13472-36-1
Synonym TSPP
Recommended Storage Store without restrictions