missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium pyrophosphate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1754630

 View more versions of this product

Catalog No. AAA1754630



Description

Description

Sodium pyrophosphate is used in water softener, detergent, emulsifying agent, metal cleaner and nutritional supplement. It acts as a buffering agent, thickening agent and dispersing agent. It also acts as a tartar control agent in toothpaste and dental floss. In addition, it is used as a chelating agent in antimicrobial studies. It is also used as a food additive in common foods such as chicken nuggets, crab meat and canned tuna.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium pyrophosphate is used in water softener, detergent, emulsifying agent, metal cleaner and nutritional supplement. It acts as a buffering agent, thickening agent and dispersing agent. It also acts as a tartar control agent in toothpaste and dental floss. In addition, it is used as a chelating agent in antimicrobial studies. It is also used as a food additive in common foods such as chicken nuggets, crab meat and canned tuna.

Solubility
Soluble in water. Insoluble in ethyl alcohol.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents and strong acids.
Specifications

Specifications

Sodium pyrophosphate
7722-88-5
Na4O7P2
MFCD00003513
tetrasodium pyrophosphate, sodium pyrophosphate, tspp, phosphotex, tetrasodium diphosphate, diphosphoric acid, tetrasodium salt, sodium diphosphate, victor tspp, natrium pyrophosphat, caswell no. 847
FQENQNTWSFEDLI-UHFFFAOYSA-J
tetrasodium;phosphonato phosphate
24403
265.9
Hygroscopic
988°C
250 g
Na4P2O7
14,9240
Soluble in water. Insoluble in ethyl alcohol.
[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
265.90
CHEBI:71240
98%
2.534

Chemical Identifiers

7722-88-5
265.90
FQENQNTWSFEDLI-UHFFFAOYSA-J
24403
tetrasodium;phosphonato phosphate
Na4O7P2
MFCD00003513
tetrasodium pyrophosphate, sodium pyrophosphate, tspp, phosphotex, tetrasodium diphosphate, diphosphoric acid, tetrasodium salt, sodium diphosphate, victor tspp, natrium pyrophosphat, caswell no. 847
CHEBI:71240
[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 231-767-1

RTECSNumber : UX7350000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only