missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Potassium Tartrate, Tetrahydrate, Powder, FCC, 99-102%, Spectrum™ Chemical

P1402, 6381-59-5, C4H4KNaO6•4H2O

$1460.34 - $4236.69

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.22
InChI Key VZOPRCCTKLAGPN-ZFJVMAEJSA-L
IUPAC Name potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
SMILES O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606754
View Documents Promotion Details
Spectrum Chemical Mfg Cor
P140225KG
2.5 kg Poly Pail
Each for $1,460.34
 
18606755
View Documents Promotion Details
Spectrum Chemical Mfg Cor
P140212KG
12 kg Poly Pail
Each for $4,236.69
 
Description

Description

Spectrum™ Chemical Sodium Potassium Tartrate, Tetrahydrate, Powder, FCC is used as a sequestrant and stabilizer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

6381-59-5
282.22
potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
C4H12KNaO10
VZOPRCCTKLAGPN-ZFJVMAEJSA-L
O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

To pass test
1
C4H12KNaO10
VZOPRCCTKLAGPN-ZFJVMAEJSA-L
potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
99 to 102%
6381-59-5
Poly Pail
2.5 kg
O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
282.22
FCC
SDS
missing translation for 'documents'

missing translation for 'documents'