missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Potassium Tartrate Tetrahydrate (Crystalline/Certified ACS), Fisher Chemical

Manufacturer:  Thermo Scientific S387500

 View more versions of this product

Catalog No. S387-500




Specifications

Specifications

Sodium Potassium Tartrate Tetrahydrate
Pass Test
437.22°C
70°C
5.5 to 8.5
99 to 102%
MFCD00150989
0.005% max. Insoluble Matter
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
282.218
282.22
99 to 102%
>100°C
Poly Bottle
White
500g
6381-59-5
C4H12KNaO10
acmc-209nif
VZOPRCCTKLAGPN-UHFFFAOYSA-L
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
2724148
Powder/Solid
Certified ACS

Chemical Identifiers

6381-59-5
282.218
VZOPRCCTKLAGPN-UHFFFAOYSA-L
2724148
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
C4H12KNaO10
MFCD00150989
acmc-209nif
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:0

SDS Documents