Please login to your online account to display your discounted pricing

Sodium Potassium Tartrate, Tetrahydrate, ACS Grade, 99.0 to 102.0%, LabChem™

$171.60 - $171.60

Chemical Identifiers

Chemical Name or Material Sodium Potassium Tartrate
CAS 6381-59-5
CAS Max % 100
Name Note Tetrahydrate
Purity Grade ACS Grade
Linear Formula NaKC4H4O6·4H2O
Formula Weight 282.23
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC247851

 
lab chem
LC247851
500g Each for $171.60
Description & Specifications

Specifications

Formula Weight 282.23
Storage Information Room Temperature
Chemical Name or Material Sodium Potassium Tartrate
EINECS Number 206-156-8
Identification Passes Test
Name Note Tetrahydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Assay 99.0 to 102.0%
Solubility Soluble in water
Linear Formula NaKC4H4O6·4H2O
CAS 6381-59-5
Decomposition Information Sodium oxide; Carbon monoxide; Carbon dioxide
Color White
Melting Point 158°C
Quantity 500g
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.